2-Chloro-6-methyl-4-nitropyridine 95%

Supplier: Ambeed
Warning

A502989-1G A502989-250MG A502989-5G A502989-25G
76818-750EA 23 USD
76818-750 76818-752 76818-754 77572-600
2-Chloro-6-methyl-4-nitropyridine 95%
2-Chloro-6-methyl-4-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MW: 172.569 g/mol
MDL Number: MFCD14582081
CAS Number: 79055-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR