(2S,4R)-1,2-Dicarbobenzyloxy-4-hydroxypyrrolidine 95%

Supplier: Ambeed
Warning

Synonyms: (2S,4R)-Dibenzyl 4-hydroxypyrrolidine-1,2-dicarboxylate

A502802-250MG A502802-5G
76818-748EA 47.75 USD
76818-748 77673-316
(2S,4R)-1,2-Dicarbobenzyloxy-4-hydroxypyrrolidine 95%
(2S,4R)-1,2-Dicarbobenzyloxy-4-hydroxypyrrolidine

Precaución: for Research Use Only

Formula: C₂₀H₂₁NO₅
MW: 355.39 g/mol
MDL Number: MFCD12407103
CAS Number: 13500-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR