4-(Chloromethyl)thiazole hydrochloride 98%

Supplier: Ambeed
Danger

A501678-10G A501678-5G
76818-740EA 15 USD
76818-740 77572-594
4-(Chloromethyl)thiazole hydrochloride 98%
4-(Chloromethyl)thiazole hydrochloride

Precaución: for Research Use Only

Formula: C₄H₄ClNS
MDL Number: MFCD02094738
CAS Number: 7709-58-2
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR