N-(4-Aminobutyl)-N-ethylisoluminol 98%

Supplier: Ambeed
Warning

Synonyms: 6-[N-(4-Aminobutyl)-N-ethylamino]-2,3-dihydro-1,4-phthalazinedione, ABEI

A500172-100MG A500172-50MG
76818-736EA 119.89 USD
76818-736 76818-738
N-(4-Aminobutyl)-N-ethylisoluminol 98%
N-(4-Aminobutyl)-N-ethylisoluminol

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₄O₂
MW: 276.34 g/mol
MDL Number: MFCD00010559
CAS Number: 66612-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR