1-Butyl-3-methylimidazolium chloride 97%

Supplier: Ambeed
Danger

Synonyms: 3-Butyl-1-methyl-1H-imidazol-3-ium chloride

A498919-100G A498919-25G A498919-500G A498919-5G
76818-726EA 29.71 USD
76818-726 76818-728 76818-730 76818-732
1-Butyl-3-methylimidazolium chloride 97%
1-Butyl-3-methylimidazolium chloride

Precaución: for Research Use Only

Formula: C₈H₁₅ClN₂
MW: 174.67 g/mol
MDL Number: MFCD03095425
CAS Number: 79917-90-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR