6-Bromo-2-fluoro-3-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A495793-5G A495793-1G
76818-716EA 250 USD
76818-716 77673-292
6-Bromo-2-fluoro-3-methylbenzaldehyde 97%
6-Bromo-2-fluoro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO
MDL Number: MFCD11520668
CAS Number: 1114809-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR