(R)-1,4-Benzodioxane-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A495025-100MG A495025-250MG A495025-5G
76818-710EA 38.2 USD
76818-710 76818-712 76818-714
(R)-1,4-Benzodioxane-2-carboxylic acid 97%
(R)-1,4-Benzodioxane-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈O₄
MW: 180.157 g/mol
MDL Number: MFCD00239407
CAS Number: 70918-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR