4-Bromo-1-butoxy-2-chlorobenzene 95+%

Supplier: Ambeed
Warning

A494838-1G A494838-250MG A494838-5G
76818-704EA 126.25 USD
76818-704 76818-706 76818-708
4-Bromo-1-butoxy-2-chlorobenzene 95+%
4-Bromo-1-butoxy-2-chlorobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrClO
MW: 263.55 g/mol
MDL Number: MFCD09907662
CAS Number: 1266253-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR