3-(Methylthio)benzotrifluoride 98%

Supplier: Ambeed
Warning

A494598-10G A494598-1G A494598-5G
76818-698EA 163 USD
76818-698 76818-700 76818-702
3-(Methylthio)benzotrifluoride 98%
3-(Methylthio)benzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₇F₃S
MDL Number: MFCD03789168
CAS Number: 328-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR