(R)-(-)-2-Amino-1-butanol 96%

Supplier: Ambeed
Danger

A494024-10G A494024-25G A494024-5G A494024-100G A494024-500G
76818-688EA 7 USD
76818-688 76818-690 76818-692 77594-550 77673-284
(R)-(-)-2-Amino-1-butanol 96%
(R)-(-)-2-Amino-1-butanol

Precaución: for Research Use Only

Formula: C₄H₁₁NO
MDL Number: MFCD00064419
CAS Number: 5856-63-3
UN: 3267
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR