4-Methylpiperazine-1-sulfonyl chloride hydrochloride 95%

Supplier: Ambeed
Danger

A492980-100MG A492980-250MG
76818-682EA 29.71 USD
76818-682 76818-684
4-Methylpiperazine-1-sulfonyl chloride hydrochloride 95%
4-Methylpiperazine-1-sulfonyl chloride hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂Cl₂N₂O₂S
MW: 235.13 g/mol
MDL Number: MFCD13186323
CAS Number: 33581-96-3
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR