4-(Thiomorpholin-4-ylsulfonyl)benzeneboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: N-Thiomorpholin-4-yl 4-boronobenzenesulphonamide, (4-(Thiomorpholinosulphonyl)phenyl)boronic acid

A491238-100MG A491238-1G A491238-250MG
76818-674EA 37.14 USD
76818-674 76818-676 76818-678
4-(Thiomorpholin-4-ylsulfonyl)benzeneboronic acid 96%
4-(Thiomorpholin-4-ylsulfonyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BNO₄S₂
MW: 287.163 g/mol
MDL Number: MFCD07363757
CAS Number: 871329-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR