4,6-Dihydrothieno[3,4-b]thiophene-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A490952-1G A490952-5G
76818-670EA 84.88 USD
76818-670 76818-672
4,6-Dihydrothieno[3,4-b]thiophene-2-carboxylic acid 97%
4,6-Dihydrothieno[3,4-b]thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆O₂S₂
MW: 186.24 g/mol
MDL Number: MFCD20642620
CAS Number: 7712-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR