(S)-(+)-3-Chloro-1,2-propanediol 98%

Supplier: Ambeed
Danger

Synonyms: (2S)-(+)-3-Chloropropane-1,2-diol, (S)-(+)-3-Chloro-1,2-dihydroxy propane

A489576-5G A489576-25G
76818-666EA 6.37 USD
76818-666 76843-234
(S)-(+)-3-Chloro-1,2-propanediol 98%
(S)-(+)-3-Chloro-1,2-propanediol

Precaución: for Research Use Only

Formula: ClCH₂CHOHCH₂OH
MW: 110.54 g/mol
MDL Number: MFCD00210270
CAS Number: 60827-45-4
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR