2-Amino-3,4-dimethoxybenzoic acid 95%

Supplier: Ambeed
Warning

A485990-100MG A485990-1G A485990-250MG
76818-654EA 28 USD
76818-654 76818-656 76818-658
2-Amino-3,4-dimethoxybenzoic acid 95%
2-Amino-3,4-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₄
MDL Number: MFCD00038792
CAS Number: 5701-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR