(2-Methylbenzo[d]oxazol-5-yl)boronic acid 98%

Supplier: Ambeed
Warning

A485978-100MG A485978-1G A485978-250MG
76818-648EA 130.5 USD
76818-648 76818-650 76818-652
(2-Methylbenzo[d]oxazol-5-yl)boronic acid 98%
(2-Methylbenzo[d]oxazol-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₃
MW: 176.96 g/mol
MDL Number: MFCD18251184
CAS Number: 279262-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR