trans-2-Aminocyclohexanol hydrochloride 98%

Supplier: Ambeed
Warning

A484581-100G A484581-10G A484581-1G A484581-250MG A484581-25G A484581-5G
76818-632EA 666 USD
76818-632 76818-634 76818-636 76818-638 76818-640 76818-642
trans-2-Aminocyclohexanol hydrochloride 98%
trans-2-Aminocyclohexanol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
MDL Number: MFCD00001490
CAS Number: 5456-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR