Methyl-1-methyl-5-oxopyrrolidine-3-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A482769-1G
A482769-25G
A482769-5G
76818-622EA
20
USD
76818-622
76818-624
76818-626
Methyl-1-methyl-5-oxopyrrolidine-3-carboxylate 98%
Methyl-1-methyl-5-oxopyrrolidine-3-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₁₁NO₃ |
MDL Number:
MFCD00015898 CAS Number: 59857-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...