4-Chloro-5-methyl-5H-pyrrolo[3,2-d]pyrimidine 98%
Supplier: Ambeed
|
Danger
|
A482165-5G
A482165-250MG
76818-618EA
581.38
USD
76818-618
77673-248
4-Chloro-5-methyl-5H-pyrrolo[3,2-d]pyrimidine 98%
4-Chloro-5-methyl-5H-pyrrolo[3,2-d]pyrimidine
Precaución: for Research Use Only
Formula:
C₇H₆ClN₃ |
MDL Number:
MFCD12924466 CAS Number: 871024-38-3 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...