2-(3-Bromo-2-methoxyphenyl)acetonitrile 97%

Supplier: Ambeed
Warning

A481233-1G A481233-5G
76818-614EA 116.7 USD
76818-614 76818-616
2-(3-Bromo-2-methoxyphenyl)acetonitrile 97%
2-(3-Bromo-2-methoxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₉H₈BrNO
MW: 226.06 g/mol
MDL Number: MFCD18391932
CAS Number: 1261602-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR