3-Chloro-2-hydroxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A480677-100MG A480677-250MG A480677-1G A480677-5G
76818-610EA 10 USD
76818-610 76818-612 77884-223 77884-224
3-Chloro-2-hydroxyphenylboronic acid 98%
3-Chloro-2-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BClO₃
MDL Number: MFCD08689551
CAS Number: 951655-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR