2,3,6-Trichloro-5-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

A477840-100MG A477840-1G A477840-250MG
76818-596EA 59 USD
76818-596 76818-598 76818-600
2,3,6-Trichloro-5-(trifluoromethyl)pyridine 95%
2,3,6-Trichloro-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆HCl₃F₃N
MDL Number: MFCD00042390
CAS Number: 80289-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR