2-Chlorobenzene-1,4-diamine sulfate(1:x) 97%

Supplier: Ambeed
Warning

A475902-100G A475902-25G A475902-500G
76818-580EA 29 USD
76818-580 76818-582 76818-584
2-Chlorobenzene-1,4-diamine sulfate(1:x) 97%
2-Chlorobenzene-1,4-diamine sulfate(1:x)

Precaución: for Research Use Only

Formula: C₆H₉ClN₂O₄S
MW: 240.66 g/mol
CAS Number: 6219-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR