D-(+)-Glucosamine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-2-deoxy-D-glucopyranose Hydrochloride, D-Glucosamine hydrochloride, (2R,3R,4S,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride
A474862-1000G
A474862-100G
A474862-25G
A474862-500G
A474862-50MG
76818-570EA
47.75
USD
76818-570
76818-572
76818-574
76818-576
76818-578
D-(+)-Glucosamine hydrochloride 98%
D-(+)-Glucosamine hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₃NO₅ MW: 215.63 g/mol |
MDL Number:
MFCD00135831 CAS Number: 66-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...