2-(3-Thienyl)-1,3-dioxolane 95% (by GC)

Supplier: Ambeed
Warning

Synonyms: 2-(Thiophen-3-yl)-1,3-dioxolane, 3-(1,3-Dioxolan-2-yl)thiophene

A474603-1G A474603-250MG A474603-25G A474603-5G
76818-562EA 12.74 USD
76818-562 76818-564 76818-566 76818-568
2-(3-Thienyl)-1,3-dioxolane 95% (by GC)
2-(3-Thienyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₇H₈O₂S
MW: 156.20 g/mol
MDL Number: MFCD06209359
CAS Number: 13250-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR