Splitomicin 95%

Supplier: Ambeed
Warning

Synonyms: 1,2-Dihydro-3H-naphtho[2,1-b]pyran-3-one, 1,2-Dihydro-3H-benzo[f]chromen-3-one

A473757-10MG A473757-1MG A473757-25MG
76818-536EA 18.04 USD
76818-536 76818-538 76818-540
Splitomicin 95%
Splitomicin

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
MDL Number: MFCD08705254
CAS Number: 5690-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR