2-Amino-5,6-dimethoxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dimethoxypyridin-2-amine

A471611-1G A471611-100MG A471611-5G
76818-532EA 154.9 USD
76818-532 77673-216 77673-218
2-Amino-5,6-dimethoxypyridine 98%
2-Amino-5,6-dimethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.167 g/mol
MDL Number: MFCD09910033
CAS Number: 127980-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR