Methyl 2-fluoro-3-hydroxybenzoate 97%

Supplier: Ambeed
Warning

A471300-100MG A471300-1G A471300-250MG A471300-5G
76818-524EA 10 USD
76818-524 76818-526 76818-528 76818-530
Methyl 2-fluoro-3-hydroxybenzoate 97%
Methyl 2-fluoro-3-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MDL Number: MFCD14698042
CAS Number: 1214324-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR