Ethyl 5-chloro-1H-pyrazole-3-carboxylate 98%

Supplier: Ambeed
Warning

A470713-100MG A470713-1G A470713-250MG A470713-5G
76818-510EA 28.65 USD
76818-510 76818-512 76818-514 76818-516
Ethyl 5-chloro-1H-pyrazole-3-carboxylate 98%
Ethyl 5-chloro-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O₂
MW: 174.58 g/mol
MDL Number: MFCD20275525
CAS Number: 1378271-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR