1-Phenylpiperidin-4-amine dihydrochloride 95%

Supplier: Ambeed
Warning

A470208-1G A470208-250MG A470208-5G
76818-504EA 138 USD
76818-504 76818-506 76818-508
1-Phenylpiperidin-4-amine dihydrochloride 95%
1-Phenylpiperidin-4-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₈Cl₂N₂
MDL Number: MFCD23135281
CAS Number: 1082662-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR