4-Fluoro-3-formyl-5-methylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-Fluoro-3-formyl-5-methylphenyl)boronic acid

A467885-1G A467885-250MG A467885-5G
76818-484EA 98.67 USD
76818-484 76818-486 76818-488
4-Fluoro-3-formyl-5-methylphenylboronic acid 98%
4-Fluoro-3-formyl-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BFO₃
MW: 181.957 g/mol
MDL Number: MFCD11111909
CAS Number: 1310384-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR