(3R,5R)-tert-Butyl 7-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)-3,5-dihydroxyheptanoate 98%

Supplier: Ambeed
Warning

A467606-1G A467606-250MG
76818-480EA 24.41 USD
76818-480 76818-482
(3R,5R)-tert-Butyl 7-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)-3,5-dihydroxyheptanoate 98%
(3R,5R)-tert-Butyl 7-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)-3,5-dihydroxyheptanoate

Precaución: for Research Use Only

Formula: C₃₇H₄₃FN₂O₅
MW: 614.74 g/mol
MDL Number: MFCD09839952
CAS Number: 134395-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR