Ethyl 3-amino-2,2-dimethylpropanoate hydrochloride 97%

Supplier: Ambeed
Warning

A467338-10G A467338-250MG A467338-25G A467338-5G
76818-472EA 152.77 USD
76818-472 76818-474 76818-476 76818-478
Ethyl 3-amino-2,2-dimethylpropanoate hydrochloride 97%
Ethyl 3-amino-2,2-dimethylpropanoate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂·ClH
MW: 181.66 g/mol
MDL Number: MFCD00102231
CAS Number: 80253-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR