(S)-α-(2-Oxiranyl)anisole 95%

Supplier: Ambeed
Danger

Synonyms: (S)-Glycidyl phenyl ether

A466894-1G A466894-25G A466894-5G
76818-466EA 23.34 USD
76818-466 76818-468 76818-470
(S)-α-(2-Oxiranyl)anisole 95%
(S)-α-(2-Oxiranyl)anisole

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.1745 g/mol
MDL Number: MFCD06659026
CAS Number: 71031-03-3
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR