H-DL-Phe(3-CN)-OH 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-(3-cyanophenyl)propanoic acid

A466366-10G A466366-1G A466366-25G A466366-5G
76818-454EA 101 USD
76818-454 76818-456 76818-458 76818-460
H-DL-Phe(3-CN)-OH 98%
H-DL-Phe(3-CN)-OH

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.2 g/mol
MDL Number: MFCD00190859
CAS Number: 63999-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR