2-Methyltetrahydro-2H-pyran-4-ol 96%

Supplier: Ambeed
Danger

A464541-1G A464541-250MG
76818-446EA 46.68 USD
76818-446 76818-448
2-Methyltetrahydro-2H-pyran-4-ol 96%
2-Methyltetrahydro-2H-pyran-4-ol

Precaución: for Research Use Only

Formula: C₆H₁₂O₂
MW: 116.158 g/mol
MDL Number: MFCD16877672
CAS Number: 89791-47-9
UN: 1987
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR