Di-tert-butyl(2',4',6'-triisopropyl-3,4,5,6-tetramethyl-[1,1'-biphenyl]-2-yl)phosphine 98%

Supplier: Ambeed
Warning

A462981-100MG A462981-1G A462981-250MG A462981-5G A462981-25G
76818-420EA 9 USD
76818-420 76818-422 76818-424 76818-426 77673-180
Di-tert-butyl(2',4',6'-triisopropyl-3,4,5,6-tetramethyl-[1,1'-biphenyl]-2-yl)phosphine 98%
Di-tert-butyl(2',4',6'-triisopropyl-3,4,5,6-tetramethyl-[1,1'-biphenyl]-2-yl)phosphine

Precaución: for Research Use Only

Formula: C₃₃H₅₃P
MW: 480.74 g/mol
MDL Number: MFCD09038436
CAS Number: 857356-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR