(R)-tert-Butyl (3-oxocyclopentyl)carbamate 95%

Supplier: Ambeed
Warning

A461162-1G A461162-250MG A461162-100MG A461162-5G
76818-414EA 376 USD
76818-414 76818-416 77884-186 77884-187
(R)-tert-Butyl (3-oxocyclopentyl)carbamate 95%
(R)-tert-Butyl (3-oxocyclopentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₃
MDL Number: MFCD18835640
CAS Number: 225641-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR