3-Amino-1-adamantanol hydrate 95% (contains < 6% H₂O)
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Amino-3-hydroxyadamantane, 3-Aminoadamantan-1-ol
A459063-10G
A459063-1G
A459063-25G
A459063-5G
A459063-100G
76818-398EA
8.49
USD
76818-398
76818-400
76818-402
77673-168
77673-166
3-Amino-1-adamantanol hydrate 95% (contains < 6% H₂O)
3-Amino-1-adamantanol hydrate
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO·xH₂O MW: 167.25 g/mol (anhy) |
MDL Number:
MFCD01821204 CAS Number: 702-82-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...