((2-Cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl)methyl)triphenylphosphonium bromide 98%

Supplier: Ambeed
Warning

A456740-250MG A456740-25G A456740-5G A456740-1G
76818-384EA 6.37 USD
76818-384 76818-386 76818-388 77673-150
((2-Cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl)methyl)triphenylphosphonium bromide 98%
((2-Cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl)methyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₃₇H₃₀BrFNP
MW: 618.51 g/mol
MDL Number: MFCD19440813
CAS Number: 154057-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR