N-(5-((Diphenylphosphoryl)methyl)-4-(4-fluorophenyl)-6-isopropylpyrimidin-2-yl)-N-methylmethanesulfonamide 97%
Supplier: Ambeed
|
Warning
|
A452016-10G
A452016-1G
A452016-25G
A452016-5G
A452016-100G
76818-350EA
10
USD
76818-350
76818-352
76818-354
76818-356
76818-358
77884-161
N-(5-((Diphenylphosphoryl)methyl)-4-(4-fluorophenyl)-6-isopropylpyrimidin-2-yl)-N-methylmethanesulfonamide 97%
N-(5-((Diphenylphosphoryl)methyl)-4-(4-fluorophenyl)-6-isopropylpyrimidin-2-yl)-N-methylmethanesulfonamide
Precaución: for Research Use Only
Formula:
C₂₈H₂₉FN₃O₃PS |
MDL Number:
MFCD09839448 CAS Number: 289042-10-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...