(3,6-Dihydro-2H-pyran-4-yl)boronic acid 98%

Supplier: Ambeed
Warning

A450510-1G A450510-250MG A450510-5G
76818-344EA 202.64 USD
76818-344 76818-346 76818-348
(3,6-Dihydro-2H-pyran-4-yl)boronic acid 98%
(3,6-Dihydro-2H-pyran-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₅H₉BO₃
MW: 127.93 g/mol
MDL Number: MFCD02179502
CAS Number: 1002127-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR