(S)-(+)-2-Methylbutyric acid 98%

Supplier: Ambeed
Danger

Synonyms: (S)-(+)-2-Methylbutanoic acid, (S)-2-Methylbutanoic acid

A450063-1G A450063-250MG A450063-5G A450063-100MG
76818-338EA 91.24 USD
76818-338 76818-340 76818-342 77673-112
(S)-(+)-2-Methylbutyric acid 98%
(S)-(+)-2-Methylbutyric acid

Precaución: for Research Use Only

Formula: C₅H₁₀O₂
MW: 102.13 g/mol
MDL Number: MFCD00063166
CAS Number: 1730-91-2
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR