Methyl 2-bromo-5-(trifluoromethyl)benzoate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-(trifluoromethyl)benzoic acid methyl ester

A449937-1G A449937-5G
76818-334EA 44.56 USD
76818-334 76818-336
Methyl 2-bromo-5-(trifluoromethyl)benzoate 98%
Methyl 2-bromo-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O₂
MW: 283.042 g/mol
MDL Number: MFCD09999461
CAS Number: 1026355-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR