3,4-Dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-benzo[b][1,4]oxazine

A449416-100MG A449416-1G A449416-250MG A449416-5G
76818-328EA 33 USD
76818-328 76818-330 76818-332 77884-152
3,4-Dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester 98%
3,4-Dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀BNO₃
MDL Number: MFCD18073255
CAS Number: 1155264-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR