DL-7-Azatryptophan monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-(7H-pyrrolo[2,3-b]pyridin-3-yl)propanoic acid, H-DL-7-Aza-Trp-OH

A448683-100MG A448683-1G A448683-250MG
76818-326EA 52 USD
76818-326 77613-386 77882-163
DL-7-Azatryptophan monohydrate 98%
DL-7-Azatryptophan monohydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₁N₃O₂.H₂O
MW: 223.24 g/mol
MDL Number: MFCD00037930
CAS Number: 7303-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR