2-Bromo-4-chloro-6-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-4-chloro-α,α,α-trifluoro-o-toluidine

A447518-100G A447518-25G A447518-5G
76818-316EA 207 USD
76818-316 76818-318 76818-320
2-Bromo-4-chloro-6-(trifluoromethyl)aniline 98%
2-Bromo-4-chloro-6-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄BrClF₃N
MW: 274.47 g/mol
MDL Number: MFCD08436083
CAS Number: 912617-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR