Benzyl-2-naphthyl ether 98%
Supplier: Ambeed
|
Warning
|
A447427-1000G
A447427-100G
A447427-500G
76818-310EA
30
USD
76818-310
76818-312
76818-314
Benzyl-2-naphthyl ether 98%
Benzyl-2-naphthyl ether
Precaución: for Research Use Only
Formula:
C₁₇H₁₄O |
MDL Number:
MFCD00089092 CAS Number: 613-62-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...