2-Amino-5-chloropyrimidine 98%

Supplier: Ambeed
Warning

A446933-100G A446933-10G A446933-25G A446933-5G A446933-500G
76818-290EA 47 USD
76818-290 76818-292 76818-294 76818-296 77613-376
2-Amino-5-chloropyrimidine 98%
2-Amino-5-chloropyrimidine

Precaución: for Research Use Only

Formula: C₄H₄ClN₃
MDL Number: MFCD00014604
CAS Number: 5428-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR