4-Bromo-7-methoxy-2,3-dihydro-1H-inden-1-one 95%

Supplier: Ambeed
Warning

A446687-100MG A446687-1G A446687-250MG A446687-5G
76818-280EA 19.1 USD
76818-280 76818-282 76818-284 76818-286
4-Bromo-7-methoxy-2,3-dihydro-1H-inden-1-one 95%
4-Bromo-7-methoxy-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₂
MW: 241.08 g/mol
MDL Number: MFCD10565602
CAS Number: 5411-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR